ASI Automated RPR Test For Syphilis, Arlington Scientific
The ASI automated RPR test kit for syphilis is a nontreponemal test for diagnostic testing, blood donor screening, and cadaveric/transplant donor screening, for use on the ASI evolution analyzer.
- Titers up to 1:2048
- Up to 24 months shelf life
- Microtiter plates included
- Run 190 tests per hour
- Made in USA
- Simple and economical to use
: Automated RPR contols sold separately.
Frequently Bought Together