American Machine & Tool Closed-Impeller Close-Coupled Centrifugal Pumps
Investment cast stainless steel (SS) pumps are built for heavy-duty, high-performance applications requiring excellent chemical resistance.
- Ideal for low-pressure OEM and chemical industry applications
- Rotate head in 90° increments for easy installation of tubing
- Self-cleaning impeller prevents clogging and reduces maintenance
These closed-Impeller close-coupled centrifugal pumps are constructed with a 316 stainless steel (SS) pump head, impeller, and shaft, along with Viton seal and o-ring.
: Includes single phase: spade connections. Three-phase: conduit box with stripped ends.
- Environmentally Preferable: