To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Club Cell Protein
- Antigen Symbol:CC16
- Clonality:Polyclonal
- Clone:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:The antibody is lyophilized in 0.05 M phosphate buffer, 0.1 M NaCl, pH 7.2. AZIDE FREE.
- Antigen Synonyms:uteroglobin|Casein kinase 1|CK1|urinary protein 1|CC10|CC16
- Storage Buffer:Add 0.2 ml of deionized water and let the lyophilized pellet dissolve completely. Slight turbidity may occur after reconstitution, which does not affect activity of the antibody. In this case clarify the solution by centrifugation.
- Shipping Temperature:At ambient temperature
- Purification:Immunoaffinity chromatography on a column with immobilized recombinant Human Club Cell Protein (RD172022100, Biovendor).
- Cat. No.:102830-376
- Supplier no.:RD181022220-01
Specifications
About this item
Quality control test, Indirect ELISA – to determine titer of the antibody , SDS PAGE – to determine purity of the antibody, and BCA - to determine quantity of the antibody.
Human Club Cell Protein (CC16, CC10 and also called uteroglobin, urinary protein 1 or Clara Cell Secretory Protein) belongs to the family of secretoglobins and is a secreted protein product of non-ciliated bronchiolar Club cells. Its function remains to be elucidated but there is convincing data suggesting its phospholipase A2 inhibitory activity as well as a number of other immunomodulatory features including inhibition of interferon gamma signaling and Th1 vs. Th2 lymphocyte regulation.
Storage temperature: The lyophilized antibody remains stable and fully active until the expiry date when stored at –20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles and store frozen at –80 °C. Reconstituted antibody can be stored at 4 °C for a limited period of time; it does not show decline in activity after one week at 4 °C.