Order Entry
Puerto Rico
ContactUsLinkComponent
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), No-Weigh™ Format, Pierce™
Irritant
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), No-Weigh™ Format, Pierce™
Catalog # 103523-940
Supplier:  Invitrogen
CAS Number:  
Irritant
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), No-Weigh™ Format, Pierce™
Catalog # 103523-940
Supplier:  Invitrogen
Supplier Number:  A35391
CAS Number:  

Specifications

  • Size:
    10 x 1 mg
  • Cat. No.:
    103523-940

Specifications

Specification Test Results

  • Cell Permeability:
    Cell-Impermeant
  • Shipping Condition:
    Wet Ice
  • Product Line:
    No-Weigh™, Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Carbodiimide
  • Spacer:
    Short
  • Form:
    Powder
  • Quantity:
    10 x 1 mg
  • Type:
    Crosslinker
  • Solubility:
    Water

Specification Test Results

About this item

 :
 :191.7 g/mol
Melting Pt:∼ 113 °C
Storage Temperature:Freezer
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:25952-53-8
Formula:C₈H₁₇N₃·HCl
MDL Number:MFCD00012503