Chorionic Gonadotropin Human
Human Chorionic Gonadotropin is produced from a sterile preparation of placental glucoprotein urine of pregnant women having a total molecular mass of 36700 Dalton.
- Urine of pregnant women
- Sterile Filtered White lyophilized (freeze-dried) powder
- The hCG was lyophilized with no additives
- It is recommended to reconstitute the lyophilized Human Chorionic Gonadotropin in sterile 18 MΩ-cm
- H₂O not less than 100 µg/ml, which can then be further diluted to other aqueous solutions
The hCG consists of 237 amino acids, a chain-92 amino acids and b chain-145 amino acids. The hCG is purified by proprietary chromatographic techniques.
Caution: Research use only.
Human Chorionic Gonadotropin is produced from a sterile preparation of placental glucoprotein urine of pregnant women having a total molecular mass of 36700 Dalton.
- Urine of pregnant women
- Sterile Filtered White lyophilized (freeze-dried) powder
- The hCG was lyophilized with no additives
- It is recommended to reconstitute the lyophilized Human Chorionic Gonadotropin in sterile 18 MΩ-cm
- H₂O not less than 100 µg/ml, which can then be further diluted to other aqueous solutions
The hCG consists of 237 amino acids, a chain-92 amino acids and b chain-145 amino acids. The hCG is purified by proprietary chromatographic techniques.
Caution: Research use only.
- Catalog No:
- 103737-128
- 103737-130
- Protein/Peptide Name:
- Chorionic Gonadotropin Human
- Chorionic Gonadotropin Human
- Protein Synonyms:
- CGH
- CGH
- Protein/Peptide Type:
- Native
- Native
- Species:
- Human
- Human
- Source:
- Urine (pregnant women)
- Urine (pregnant women)
- Conjugation:
- Unconjugated
- Unconjugated
- Biological Activity:
- The activity was found to be 5625 IU/mg (may vary slightly with each lot)
- The activity was found to be 5625 IU/mg (may vary slightly with each lot)
- Formulation:
- The hCG was lyophilized with no additives
- The hCG was lyophilized with no additives
- Reconstitution Instructions:
- It is recommended to reconstitute the lyophilized Human Chorionic Gonadotropin in sterile 18 MΩ-cm H₂O not less than 100 µg/ml, which can then be further diluted to other aqueous solutions.
- It is recommended to reconstitute the lyophilized Human Chorionic Gonadotropin in sterile 18 MΩ-cm H₂O not less than 100 µg/ml, which can then be further diluted to other aqueous solutions.
- Storage Conditions:
- Lyophilized hCG although stable at room temperature for 3 weeks, should be stored desiccated below −18 °C. Upon reconstitution CG-beta should be stored at 4 °C between 2 - 7 days and for future use below −18 °C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA). Please prevent freeze-thaw cycles
- Lyophilized hCG although stable at room temperature for 3 weeks, should be stored desiccated below −18 °C. Upon reconstitution CG-beta should be stored at 4 °C between 2 - 7 days and for future use below −18 °C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA). Please prevent freeze-thaw cycles
Product Family Options
- Size
- : RP17825005Marketsource
Specifications
Cat. No.103737-128Protein/Peptide NameChorionic Gonadotropin HumanProtein SynonymsCGHProtein/Peptide TypeNativeSpeciesHumanSourceUrine (pregnant women)ConjugationUnconjugatedBiological ActivityThe activity was found to be 5625 IU/mg (may vary slightly with each lot)FormulationThe hCG was lyophilized with no additivesReconstitution InstructionsIt is recommended to reconstitute the lyophilized Human Chorionic Gonadotropin in sterile 18 MΩ-cm H₂O not less than 100 µg/ml, which can then be further diluted to other aqueous solutions.Storage ConditionsLyophilized hCG although stable at room temperature for 3 weeks, should be stored desiccated below −18 °C. Upon reconstitution CG-beta should be stored at 4 °C between 2 - 7 days and for future use below −18 °C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA). Please prevent freeze-thaw cyclesMarketsource - : RP17825010Marketsource
Specifications
Cat. No.103737-130Protein/Peptide NameChorionic Gonadotropin HumanProtein SynonymsCGHProtein/Peptide TypeNativeSpeciesHumanSourceUrine (pregnant women)ConjugationUnconjugatedBiological ActivityThe activity was found to be 5625 IU/mg (may vary slightly with each lot)FormulationThe hCG was lyophilized with no additivesReconstitution InstructionsIt is recommended to reconstitute the lyophilized Human Chorionic Gonadotropin in sterile 18 MΩ-cm H₂O not less than 100 µg/ml, which can then be further diluted to other aqueous solutions.Storage ConditionsLyophilized hCG although stable at room temperature for 3 weeks, should be stored desiccated below −18 °C. Upon reconstitution CG-beta should be stored at 4 °C between 2 - 7 days and for future use below −18 °C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA). Please prevent freeze-thaw cyclesMarketsource
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



