Order Entry
Puerto Rico
ContactUsLinkComponent
Mammaglobin A (Fc Chimera) Human HEK293, 0.1 mg
Mammaglobin A (Fc Chimera) Human HEK293, 0.1 mg
Catalog # 103739-808
Supplier:  BioVendor
CAS Number:  
undefined
Mammaglobin A (Fc Chimera) Human HEK293, 0.1 mg
Catalog # 103739-808
Supplier:  BioVendor
Supplier Number:  RD172188100HEK
CAS Number:  

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.1 mg
  • Storage Conditions:
    Store the lyophilized protein at −80 °C. Lyophilized protein remains stable until the expiry date when stored at −80 °C. Aliquot reconstituted protein to avoid repeated freezing/thawing cycles and store at −80 °C for long term storage. Reconstituted protein can be stored at 4 °C for 5 days
  • Endotoxin Content:
    <0.1 EU/μg
  • Reconstitution Instructions:
    Add deionized water to prepare a working stock solution of approximately 0.5 mg/ml and let the lyophilized pellet dissolve completely. Filter sterilize your culture media/working solutions containing this non-sterile product before using in cell culture.
  • Protein Synonyms:
    Secretoglobin family 2A member 2|Mammaglobin-1|SCGB2A2|MGB1|UGB2
  • Protein/Peptide Name:
    Mammaglobin A (Fc Chimera) Human HEK293
  • Purity:
    >95%
  • Molecular Weight:
    36.3 kDa (calculated)
  • Sequence:
    GSGCPLLENVISKTINPQVSKTEYKELLQEFIDDNATTNAIDELKECFLNQTDETLSNVEVFMQLIYDSSLCDLFKLENLYFQGPKSCDKTHTCPPCPAPELLGGPSVFLFPPKPKDTLMISRTPEVTCVVVDVSHEDPEVKFNWYVDGVEVHNAKTKPREEQYNSTYRVVSVLTVLHQDWLNGKEYKCKVSNKALPAPIEKTISKAKGQPREPQVYTLPPSRDELTKNQVSLTCLVKGFYPSDIAVEWESNGQPENNYKTTPPVLDSDGSFFLYSKLTVDKSRWQQGNVFSCSVMHEALHNHYTQKSLSLSPGKHHHHHH
  • Endotoxin Level:
    <0.1 EU/μg
  • Formulation:
    Filtered (0.4 μm) and lyophilized from 0.5 mg/ml solution in phosphate buffered saline, 5% w/v trehalose pH 7.4
  • Shipping Temperature:
    At ambient temperature
  • Tested Applications:
    Western blotting, ELISA, Cell culture and/or animal studies
  • Cat. No.:
    103739-808

Specifications

About this item

Total 321 AA. MW: 36.3 kDa (calculated). UniProtKB acc. No. Q13296 (Gly19–Phe93). C-terminal linker (2 extra AA), TEV site (7 AA), Hu-IgG1 fragment (Pro100-Lys330, 231 AA) and C-terminal His-tag (6 AA). Protein identity confirmed by LC-MS/MS.

  • Quality control test
  • BCA to determine quantity of the protein
  • SDS PAGE to determine purity of the protein
  • LAL to determine quantity of endotoxin
Caution: Research use only.