
5-Norbornene-2-carboxylic acid 98% (mixture of isomers)
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:120-74-1
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00085356
Boiling Pt:136…138 °C (14 mmHg)
Density:1.129 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:138.17 g/mol
:GHS05
Specification Test Results
- Appearance:Colorless to light yellow Liquid or solid
- Infrared spectrum:Conforms
- Refractive index:1.4925 to 1.4965 (20°C, 589 nm)
- GC:≥97.5 % (total of isomers)
- Titration with NaOH:97.5 to 102.5 %