(S)-(-)-3-Hydroxy-3-phenylpropionic acid 99%
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:36567-72-3
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00145262
:166.18 g/mol
Melting Pt:116…119 °C
Storage Temperature:Refrigerator
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:White to off-white Crystals or needles
- Infrared spectrum:Conforms
- Melting point:116 to 119 °C
- Specific optical rotation:-21.5° to -23.5° (20 °C, 589 nm) (c=4, CH3OH)
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together