Order Entry
Puerto Rico
ContactUsLinkComponent
3-Amino-6-bromopyrazine-2-carboxylic acid 97%
3-Amino-6-bromopyrazine-2-carboxylic acid 97%
Catalog # TS45742-0050
CAS Number:  
undefined
3-Amino-6-bromopyrazine-2-carboxylic acid 97%
Catalog # TS45742-0050
Supplier Number:  45742-0050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    5 g
  • Cat. No.:
    TS45742-0050

Specifications

Specification Test Results

  • Appearance:
    Off-white to brown Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :218.01 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:486424-37-7
Formula:C₅H₄BrN₃O₂
MDL Number:MFCD08275681