



1,5-Cyclooctadiene 99+% stabilized, AcroSeal™, purified by redistillation
Catalog # TS11141-1000
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle, AcroSeal®
- Size:100 ml
- Cat. No.:TS11141-1000
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4920 to 1.4950 (20°C, 589 nm)
- GC:≥99.0 %
- Stabilizer:100 to 200 ppm octadecyl 3-(3',5'-di-tert-butyl-4'-hydroxyphenyl) propionate
Specification Test Results
About this item
:
:108.18 g/mol
Boiling Pt:151 °C (101.3kPa)
Melting Pt:–69.5 °C
Density:0.88 g/cm³
Flash Pt:38 °C
Storage Temperature:Ambient
UN:2520
ADR:3,III
:GHS07
:GHS02
:GHS08
:GHS09
CAS Number:111-78-4
Formula:C₈H₁₂
MDL Number:MFCD00001752