Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Propionic acid ACS

Propionic acid ACS

Corrosive
Flammable
Irritant
Propionic acid ACS
Certificates
Propionic acid ACS

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Boiling Pt:141 °C (760 mmHg)
  • Melting Pt:–22 °C
  • Density:0.99 g/cm³
  • Flash Pt:51 °C
  • Storage Temperature:Ambient
  • UN:3463
  • ADR:8,II
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.15 %
  • Color scale:
    ≤20 APHA
  • Residue after evaporation:
    ≤0.01 %
  • Titration with NaOH:
    ≥99.5 %
  • Heavy metals (as Pb):
    ≤0.001 %
  • Carbonyl compounds:
    ≤0.002 %
  • Oxidisable substance (HCOOH):
    ≤0.1 %
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    100 ml
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $56.70
    $56.70
    Each (100ml)
       $56.70

    Supplier Number: 44723-1000
  • Glass bottle
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $104.00
    $104.00
    Each (500ml)
       $104.00

    Supplier Number: 44723-5000

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.