
Citric acid monohydrate for HPLC
Catalog # TS46454-1000
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:100 g
- Cat. No.:TS46454-1000
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- UV:(1M in water)
- Titration with NaOH:≤99.75 %
Specification Test Results
About this item
:
:210.14 g/mol
Melting Pt:135…152 °C
Flash Pt:174 °C
:GHS07
Storage Temperature:Ambient
CAS Number:5949-29-1
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972