
Adamantane-1-carboxylic acid 99%
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:828-51-3
:
Formula:C₁₁H₁₆O₂
MDL Number:MFCD00074720
:180.25 g/mol
Melting Pt:172…176 °C
EINECS:212-584-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to light beige Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:172 to 176 °C
- Titration with NaOH:98.5 to 101.5 %