Order Entry
Puerto Rico
ContactUsLinkComponent
Citric acid, anhydrous for molecular biology
Irritant
Citric acid, anhydrous for molecular biology
Catalog # TS32986-0010
CAS Number:  
undefined
Irritant
Citric acid, anhydrous for molecular biology
Catalog # TS32986-0010
Supplier Number:  32986-0010
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    1 kg
  • Cat. No.:
    TS32986-0010

Specifications

Specification Test Results

  • Appearance:
    White Crystalline solid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.5 % (K.F.)
  • Titration with NaOH:
    ≥99 %
  • Dnase,Rnase,Protease activity:
    none detected
  • Carboniz. sub. by hot H2SO4:
    (as per ACS) passes test
  • Chloride (Cl):
    ≤10 ppm
  • Oxalate (C2O4):
    ≤500 ppm
  • Insoluble matter:
    ≤0.005 %

Specification Test Results

About this item

 :
 :192.13 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:153 °C
Flash Pt:100 °C
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
 :GHS07
CAS Number:77-92-9
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669