Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Propionic acid 99+%, extra pure

Propionic acid 99+%, extra pure

Corrosive
Flammable
Irritant
Propionic acid 99+%, extra pure
Certificates
Propionic acid 99+%, extra pure

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Merck Index:13,07917
  • Boiling Pt:141 °C (1013 hPa)
  • Melting Pt:–22 °C
  • Density:0.99 g/cm³
  • Flash Pt:51 °C
  • Storage Temperature:Ambient
  • EINECS:201-176-3
  • UN:3463
  • ADR:8,II
  • REACH:01-2119486971-24
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3850 to 1.3870 (20°C, 589 nm)
  • Water:
    ≤0.1 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    Each (1l)
    Each (1l) - $78.90
    $78.90
    Each (1l)
       $78.90

    Supplier Number: 22013-0010
  • Glass bottle
    2.5 L
    Log in for availability
    Log in for availability
    Each (2,500ml)
    Each (2,500ml) - $166.00
    $166.00
    Each (2,500ml)
       $166.00

    Supplier Number: 22013-0025

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.