Order Entry
Puerto Rico
ContactUsLinkComponent
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate), No-Weigh™ Format, Pierce™
Irritant
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate), No-Weigh™ Format, Pierce™
Catalog # 103523-926
Supplier:  Invitrogen
CAS Number:  
Irritant
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate), No-Weigh™ Format, Pierce™
Catalog # 103523-926
Supplier:  Invitrogen
Supplier Number:  A35394
CAS Number:  

Specifications

  • Size:
    10 x 1 mg
  • Cat. No.:
    103523-926

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Maleimide, NHS Ester
  • Spacer:
    Short
  • Form:
    Powder
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO

Specification Test Results

About this item

 :
 :334.33 g/mol
Melting Pt:179 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:64987-85-5
Formula:C₁₆H₁₈N₂O₆
MDL Number:MFCD00009634