Order Entry
Puerto Rico
ContactUsLinkComponent
Spectrophotometer UV5Nano, UV/VIS Excellence, METTLER TOLEDO®
Spectrophotometer UV5Nano, UV/VIS Excellence, METTLER TOLEDO®
Catalog # 10811-204
Supplier:  Mettler Toledo
CSA Certified
Product available on GSA Advantage®
UL Listed
undefined
Spectrophotometer UV5Nano, UV/VIS Excellence, METTLER TOLEDO®
Catalog # 10811-204
Supplier:  Mettler Toledo
Supplier Number:  30254729
CSA Certified
Product available on GSA Advantage®
UL Listed

Specifications

  • Model:
    UV5Nano Excellence UV/VIS Spectrophotometer
  • Resolution:
    >1.7 (Toluene in Hexane)
  • Path Length:
    0.1 mm; 1.0 mm
  • Wavelength Range:
    190 - 1100 nm
  • Wavelength Accuracy:
    +/-0.9 nm
  • Photometric Accuracy:
    ±0.006A (K2Cr2O7)
  • Cat. No.:
    10811-204
  • Supplier no.:
    30254729

Specifications

About this item

The Spectrophotometer UV5Nano is ideal for life science applications such as concentration and purity determinations of DNA, RNA and protein samples using A280, A260, A260/A280, Bradford and BCA protein assays and more.

  • Micro-volume and Kinetics measurements