Order Entry
Puerto Rico
ContactUsLinkComponent
Anhydrotetracycline Hydrochloride, can be used as a Secondary Standard, Thermo Scientific
Anhydrotetracycline Hydrochloride, can be used as a Secondary Standard, Thermo Scientific
Catalog # TS23313-5000
CAS Number:  
undefined
Anhydrotetracycline Hydrochloride, can be used as a Secondary Standard, Thermo Scientific
Catalog # TS23313-5000
Supplier Number:  23313-5000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • CAS No.:
    13803-65-1
  • Description:
    Anhydrotetracycline hydrochloride
  • Size:
    500 mg
  • Formula:
    C₂₂H₂₂N₂O₇.HCl
  • Cat. No.:
    TS23313-5000

Specifications

About this item

Purity: ≥80% (as is, calculated).

Melting Point: 223 °C (D)
MDL No.: MFCD00151453
GHS Symbols: GHS07, GHS08,
Signal word: Warning

Caution: Storage Temperature: Ambient