Tris acetate 99%
Supplier: Thermo Scientific Chemicals
About this item
Synonyms: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
- CAS Number:6850-28-8
- Formula:NH₂C(CH₂OH)₃·CH₃COOH
- MDL Number:MFCD00038951
- Molecular Weight:181.1870 g/mol
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
- CAS Number:6850-28-8
- Formula:NH₂C(CH₂OH)₃·CH₃COOH
- MDL Number:MFCD00038951
- Molecular Weight:181.1870 g/mol
- Storage Temperature:Ambient
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder
- Water:≤1.0 %
- Solubility:(1 g/ml in water) Clear colorless
- UV:(50 % in water)
- Titration with NaOH:≥98.5 %
- Heavy metals (as Pb):≤5 ppm
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: 43450-0250
- Supplier Number: 43450-1000
- Supplier Number: 43450-5000
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



