Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Hydrocortisone 98%

Hydrocortisone 98%

Health
Hydrocortisone 98%
Certificates
Hydrocortisone 98%

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:50-23-7
  • Formula:C₂₁H₃₀O₅
  • MDL Number:MFCD00011654
  • Molecular Weight:362.47 g/mol
  • Melting Pt:212…217 °C
  • Storage Temperature:Ambient
  • EINECS:200-020-1
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    212 to 217 °C
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +150° to +156° (20 °C, 589 nm) (C=1, dioxane)
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $42.10
    $42.10
    Each (1g)
       $42.10

    Supplier Number: 35245-0010
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $135.00
    $135.00
    Each (5g)
       $135.00

    Supplier Number: 35245-0050
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $463.00
    $463.00
    Each (25g)
       $463.00

    Supplier Number: 35245-0250
  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $1,220.00
    $1,220.00
    Each (100g)
       $1,220.00

    Supplier Number: 35245-1000

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.