DL-Lysine monohydrate 97%
Catalog # TS36565-0250
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle for solids
- Size:25 g
- Cat. No.:TS36565-0250
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:7 % to 12 % (105°C, 2 hrs)
- Titration with HClO4:≥96.0 %
Specification Test Results
About this item
:
:164.2 g/mol
EINECS:200-740-6
CAS Number:885701-25-7
Formula:C₆H₁₄N₂O₂·1H₂O
MDL Number:MFCD00151036