Order Entry
Puerto Rico
ContactUsLinkComponent
SARS-CoV-2 NP Ab ELISA, BioVendor
SARS-CoV-2 NP Ab ELISA, BioVendor
Catalog # 103853-658
Supplier:  BioVendor
CAS Number:  
SARS-CoV-2 NP Ab ELISA, BioVendor
Catalog # 103853-658
Supplier:  BioVendor
Supplier Number:  41A225R
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Direct
  • Conjugate ELISA:
    HRP
  • Format:
    Pre-coated
  • Host:
    Human
  • Primary antibody reactivity:
    Human
  • Target protein:
    anti-SARS-CoV-2 nucleocapsid protein
  • Description:
    SARS-CoV-2 NP Ab ELISA
  • Size:
    1 kit
  • Sample Type:
    Serum, Plasma
  • Detection Method:
    Colorimetric
  • Shelf Life:
    See expiration date
  • Storage Temperature:
    2…8 °C
  • Sample Volume:
    1 µl
  • Regulatory Status:
    RUO
  • Cat. No.:
    103853-658

Specifications

About this item

96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of anti-SARS-CoV-2 nucleocapsid protein.

  • Good for research into animal studies, diabetology - other relevant products, energy metabolism and body weight regulation
  • Raised in rabbits by immunization with the recombinant Human Resistin
  • 0.1 mg (determined by BCA method, BSA was used as a standard) lyophilized in 0.05 M phosphate buffer, 0.1 M NaCl, pH 7.2. AZIDE FREE

Human resistin contains 108 amino acids as a prepeptide, and its hydrofobic signal peptide is cleaved before its secretion. Resistin circulates in human blood as a dimeric protein consisting of two 92 amino acid polypeptides, which are disulfide-linked via Cys26. Resistin may be an important link between obesity and insulin resistance. Mouse resistin, specifically produced and secreted by adipocyte, acts on skeletal muscle myocytes, hepatocytes and adipocytes themselves so that it reduces their sensitivity to insulin.

For recunstitution add 0.1 ml of deionized water and let the lyophilized pellet dissolve completely. Slight turbidity may occur after reconstitution, which does not affect activity of the antibody. In this case clarify the solution by centrifugation.

The lyophilized antibody remains stable and fully active until the expiry date when stored at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles and store frozen at −80 °C. Reconstituted antibody can be stored at 4 °C for a limited period of time; it does not show decline in activity after one week at 4 °C.

Delivery: Purified by Immunoaffinity chromatography on a column with immobilized recombinant Human Resistin
Caution: This is intended for research use only.