Sentinel XT® Starter Kit, ILC Dover
Catalog # 76533-610
Supplier: ILC DOVER LP
Specifications
- Description:Sentinel XT® starter kit
- Cat. No.:76533-610
Specifications
About this item
The Sentinel XT® blower, when paired with ILC Dover’s lithium polymer battery, weighs just 2.2 pounds, yet delivers 8+ CFM airflow, exceeding NIOSH requirements.
- Proven technology with 10+ year track record of reliability
- Brushless motor design for long life and continued quiet operation
- Robust design with secondary impact protection chamber for air handling unit
The Sentinel XT® Starter kit includes the Sentinel XT® Blower, LiPo rechargeable battery, flow meter, charging cradle, and Quick Loc belt.