ContactUsLinkComponent
Sentinel XT® Clear Plus Full Hood, ILC Dover
Sentinel XT® Clear Plus Full Hood, ILC Dover
Catalog # 76533-606
Supplier:  ILC DOVER LP
undefined
Sentinel XT® Clear Plus Full Hood, ILC Dover
Catalog # 76533-606
Supplier:  ILC DOVER LP
Supplier Number:  S-3324-30
Specifications
  • Description:
    Sentinel XT® clear plus full hood
  • Size:
    Universal
  • Cat. No.:
    76533-606
Specifications
About this item
The Sentinel XT® Clear Plus Full Hood is a superior solution when compared to other PAPR masks and facepieces.
  • Ideal for use in the Pharmaceutical industry
  • Designed for use with the Sentinel XT® Blower system
  • 320 degree field of view
  • No fit test required
  • Save time and storage space with just one size
  • APF of 1000
  • Latex-free
With an APF of 1000, the Clear Plus Full Hood offers a 320 degree field of view, allowing the wearer to have facial hair and/or a head covering, and reducing feelings of claustrophobia. Wrap-around straps allow the outer bib to secure to protective clothing.