J&W CP-Wax 57 CB Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
Unique selectivity especially for the analysis of alcohols in the brewing and wine/spirits industry.
- Unique high-polarity bonded wax column
- Excellent inertness for optimum peak shape of alcohols and glycols
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
The high inertness of this column offers excellent peak shapes for these very polar compounds, ensuring high precision. The 0.15 mm Int.Ø version offers a significant gain in analysis speed for high-speed throughput.
Specifications
- Catalog No:
- AGCP97638
- AGCP97713
- AGCP97763
- AGCP97773
- AGCP97753
- AGCP97723
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase