Order Entry
Puerto Rico
ContactUsLinkComponent
Bio-Monolith, Agilent Technologies
Bio-Monolith, Agilent Technologies
Catalog # AG5069-3637
Bio-Monolith, Agilent Technologies
Catalog # AG5069-3637
Supplier Number:  5069-3637

Specifications

  • Column Format:
    Analytical
  • Ø int.:
    5.20 mm
  • Max. Pressure:
    150 bar
  • Description:
    Bio-Monolith SO3
  • Length:
    5 mm
  • Wetted Materials:
    20% Ethanol
  • pH Range:
    2-11
  • Temperature Limit:
    40 °C
  • Column hardware:
    Stainless Steel
  • Cat. No.:
    AG5069-3637
  • Packing material:
    Monolith
  • Phase:
    SCX

Specifications

About this item

Monolithic IEX for rapid separation of antibodies, plasmid DNA, viruses, phages and other macromolcules.

  • Polymer-based, monolith HPLC columns designed for macro biomolecule separations
  • Flow-rate independent separations
  • No diffusion, no pores and no void volume make transport between mobile and stationary phase very rapid
  • Monolith disk is 5.2×4.95 mm (100 μL column volume) with continuous channels, eliminating diffusion mass transfer
  • Extremely fast separations speed up method development time and decrease costs
  • locking in method parameters

Agilent Bio-Monolith HPLC columns provide high resolution and rapid separations of antibodies (IgG, IgM), plasmid DNA, viruses, phages and other macro biomolecules. The product family offers strong cation exchange, strong and weak anion exchange, and Protein A phases.

These columns deliver flow rate-independent separation, speeding up method development time and decreasing costs. Bio-Monolith HPLC columns are compatible with HPLC and preparative LC systems, including Agilent 1100 and 1200 Infinity Series.