J&W Select Al2O3 MAPD Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
An aluminum oxide PLOT column that is stable to 200 °C for separating reactive hydrocarbons.
- Aluminum oxide PLOT column for the analysis of reactive hydrocarbons such as methyl acetylene and propadiene (MAPD)
- Optimized to improve sensitivity and response
- Faster run time improves operating efficiency
- Two-fold higher response for MAPD, especially important when running impurity analyses
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W Select Al₂O₃ MAPD is an aluminum oxide PLOT column for the analysis of reactive hydrocarbons, specially targeted towards methyl acetylene and propadiene (MAPD). The column is stable up to 200 °C for hydrocarbons up to C10. Adsorption and nonstable response for reactive hydrocarbons are greatly improved; the column delivers up to a two-fold higher response for MAPD, which is particularly important when running impurity analyses.
Specifications
- Catalog No:
- AGCP7431
- AGCP7432
- Description:
- GC Column, 7 inch cage
- GC Column, 7 inch cage