J&W CP-Silica PLOT Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Certificates
About this item
PLOT column ideal for COS in ethylene, freons/CFCs, hydrocarbons, propylene, and sulfur gases.
- No influence of water on retention times
- Elution of CO₂ and sulfur gases at ppm levels
- Separates cyclopropane from propylene
- No negative influence on retention or peak shape when water is present in the sample
- Inert surface preparation avoids decomposition of pentadienes and freons
- Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
- Performance Summary shipped with each column
Agilent J&W CP-Silica PLOT brings the benefits of capillary PLOT columns, higher efficiency and faster analysis time to many applications that previously could only be done using packed columns. It is ideal for COS in ethylene, freons/CFCs, hydrocarbons, propylene, and sulfur gases. The column offers high selectivity of C1 to C4 isomers in the presence of water, without the influence of water on retention times. CP-SilicaPLOT elutes CO₂ and sulfur gases at ppm levels and separates cyclopropane from propylene, with no decomposition of pentadienes or CFCs.
Frequently Bought Together