Sentinel XL® HP Clear Full Hood System, ILC Dover
Catalog # 76533-552
Supplier: ILC DOVER LP
Specifications
- Description:Sentinel XL® HP Clear full hood system
- Size:Universal
- Environmentally Preferable:
- Cat. No.:76533-552
Specifications
About this item
Designed for use with Sentinel XL® blower, the Sentinel XL® HP clear full hood system is ideal for use in healthcare settings.
- No fit test required
- Save time and storage space with just one size
- APF of 1000
- Available with HiMH rechargeable battery or Alkaline battery pack
- Available with or without duffel bag
- Latex-free
Ideal for HealthCare settings, the Sentinel XL® HP Clear Full Hood System includes the S-3101 Clear Hood, 33" breathing tube, Sentinel XL® Blower, flow meter, plug, NiMH rechargeable battery, charging cord, Quick Loc belt, 2 HEPA filters, and carrying tote.