ContactUsLinkComponent
Sentinel XL® HP Clear Half Hood System, ILC Dover
Sentinel XL® HP Clear Half Hood System, ILC Dover
Catalog # 76533-544
Supplier:  ILC DOVER LP
undefined
Sentinel XL® HP Clear Half Hood System, ILC Dover
Catalog # 76533-544
Supplier:  ILC DOVER LP
Supplier Number:  S-3000-01
Specifications
  • Description:
    Sentinel XL® HP Clear half hood system
  • Size:
    Universal
  • Cat. No.:
    76533-544
Specifications
About this item
Designed for use with Sentinel XL® blower, the Sentinel XL® HP clear half hood system is ideal for use in healthcare settings.
  • No fit test required
  • APF of 25
  • Can be used easily with a stethoscope
  • Available with or without duffel bag
  • Available with HiMH rechargeable battery or alkaline battery pack
  • Latex-free
Ideal for healthcare settings, the Sentinel XL® HP Clear Half Hood System includes the S-4001 clear half hood, 33" breathing tube, Sentinel XL Blower, flow meter, plug, NiMH rechargeable battery or alkaline battery pack, charging cord, Quick Loc belt, 2 HEPA filters, and carrying tote.