Nordiazepam (CRM), Cayman Chemical
76544-680
:
- Concentration:1 mg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- CAS No.:1088-11-5
- Description:Nordiazepam (CRM)
- Size:1 mg
- Synonym(s):Dealkylprazepam; Demethyldiazepam; Desmethyldiazepam; Nordazepam; Norprazepam; NSC 46078; NSC 631619; Ro 5-2180
- Formula:C₁₅H₁₁ClN₂O
- Storage Temperature:–20 °C
- Description:Nordiazepam
- Cat. No.:76544-680
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard