Phenazepam (CRM), Cayman Chemical
Catalog # 76544-676
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Concentration:1 mg/ml
- Matrix:Acetonitrile
- Pack type:Glass ampule
- CAS No.:51753-57-2
- Description:Phenazepam (CRM)
- Size:1 mg
- Synonym(s):BD 98; Fenazepam
- Formula:C₁₅H₁₀BrClN₂O
- Storage Temperature:–20 °C
- Description:Phenazepam
- Cat. No.:76544-676
Specifications
About this item
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard