(±)-Methamphetamine-d5 (hydrochloride) (CRM), Cayman Chemical
76544-670
:
- Concentration:1 mg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- CAS No.:2747917-87-7
- Description:(±)-Methamphetamine-d5 (hydrochloride) (CRM)
- Size:1 mg
- Formula:C₁₀H₁₀D₅N · HCl
- Storage Temperature:–20 °C
- Cat. No.:76544-670
A Certified Reference Material
Synonyms:DL-Methamphetamine-d5