Dextromethorphan (CRM), Cayman Chemical
Catalog # 76544-628
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Concentration:1 mg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- CAS No.:125-71-3
- Description:Dextromethorphan (CRM)
- Size:1 mg
- Formula:C₁₈H₂₅NO
- Storage Temperature:–20 °C
- Cat. No.:76544-628
Specifications
About this item
A Certified Reference Material
Synonyms:DXM;D-Methorphan