Order Entry
Puerto Rico
ContactUsLinkComponent
Acetyl norfentanyl (hydrochloride) (CRM), Cayman Chemical
Acetyl norfentanyl (hydrochloride) (CRM), Cayman Chemical
Certificates
undefined
Acetyl norfentanyl (hydrochloride) (CRM), Cayman Chemical

Specifications

  • CAS No.:
    22352-82-5
  • Formula:
    C₁₃H₁₈N₂O·HCl
  • Storage Temperature:
    –20 °C
  • Description:
    Acetyl norfentanyl (hydrochloride)

Specifications

Frequently Bought Together