Order Entry
Puerto Rico
ContactUsLinkComponent
Acetyl fentanyl-d5 (hydrochloride) (CRM), Cayman Chemical
Acetyl fentanyl-d5 (hydrochloride) (CRM), Cayman Chemical
Certificates
Acetyl fentanyl-d5 (hydrochloride) (CRM), Cayman Chemical

Specifications

  • Synonym(s):
    Desmethyl fentanyl-d5
  • Formula:
    C₂₁H₂₁D₅N₂O·HCl
  • Storage Temperature:
    –20 °C
  • Description:
    Acetyl fentanyl-d5 (hydrochloride)

Specifications