Order Entry
Puerto Rico
ContactUsLinkComponent
 
Acetyl fentanyl (hydrochloride) (CRM), Cayman Chemical
 
Acetyl fentanyl (hydrochloride) (CRM), Cayman Chemical

 

  • CAS No.:
    117332-89-5
  • Synonym(s):
    Desmethyl fentanyl; MCV 4848; NIH 10485
  • Formula:
    C₂₁H₂₆N₂O·HCl
  • Storage Temperature:
    –20 °C
  • Description:
    Acetyl fentanyl (hydrochloride)

 

Frequently Bought Together