(6aR,9S)-Δ10-THC (CRM), Cayman Chemical
Catalog # 76544-574
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Concentration:1 mg/ml
- Matrix:Acetonitrile
- Pack type:Glass ampule
- CAS No.:95588-87-7
- Description:(6aR,9S)-Δ10-THC (CRM)
- Size:1 mg
- Synonym(s):(1S,4R)-Δ2-THC; (6aR,9S)-Δ10-Tetrahydrocannabinol
- Formula:C₂₁H₃₀O₂
- Storage Temperature:–20 °C
- Description:(6aR,9S)-Δ10-THC
- Cat. No.:76544-574
Specifications
About this item
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard