(6aR,9R)-Δ10-THC (CRM), Cayman Chemical
76544-572
:
- Concentration:1 mg/ml
- Matrix:Acetonitrile
- Pack type:Glass ampule
- CAS No.:95543-62-7
- Description:(6aR,9R)-Δ10-THC (CRM)
- Size:1 mg
- Synonym(s):(6aR,9R)-Δ10-Tetrahydrocannabinol; (1R,4R)-Δ2-THC
- Formula:C₂₁H₃₀O₂
- Storage Temperature:–20 °C
- Description:(6aR,9R)-Δ10-THC
- Cat. No.:76544-572
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard