Phytocannabinoid Mixture 6 (CRM), Cayman Chemical
Catalog # 76544-350
Supplier: Cayman Chemical Company
CAS Number:
Specifications
- Concentration:500 µg/ml of each compound
- Matrix:Acetonitrile
- Pack type:Glass ampule
- Description:Phytocannabinoid Mixture 6 (CRM)
- Size:1 mg
- Storage Temperature:–20 °C
- Cat. No.:76544-350
Specifications
About this item
A mixture of phytocannabinoids in an acetonitrile solution intended for HPLC-UV or LC- or GC-MS
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard