N-Desmethylflunitrazepam (CRM), Cayman Chemical
76544-332
:
- Concentration:1 mg/ml
- Matrix:Methanol
- Pack type:Glass ampule
- CAS No.:2558-30-7
- Description:N-Desmethylflunitrazepam (CRM)
- Size:1 mg
- Synonym(s):Desmethylflunitrazepam; Norflunitrazepam; Ro 5-4435
- Formula:C₁₅H₁₀FN₃O₃
- Storage Temperature:–20 °C
- Description:N-Desmethylflunitrazepam
- Cat. No.:76544-332
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard