Order Entry
Puerto Rico
ContactUsLinkComponent
α-Pyrrolidinopentiophenone (hydrochloride) (CRM), Cayman Chemical
α-Pyrrolidinopentiophenone (hydrochloride) (CRM), Cayman Chemical
Catalog # 76544-310
CAS Number:  
α-Pyrrolidinopentiophenone (hydrochloride) (CRM), Cayman Chemical
Catalog # 76544-310
Supplier Number:  22808-1
CAS Number:  

Specifications

  • Concentration:
    1 mg/ml
  • Matrix:
    Methanol
  • Pack type:
    Glass ampule
  • CAS No.:
    5485-65-4
  • Description:
    α-Pyrrolidinopentiophenone (hydrochloride) (CRM)
  • Size:
    1 mg
  • Synonym(s):
    O-2387; α-PVP; α-Pyrrolidinovalerophenone; 2-(1-pyrrolidinyl)-Valerophenone
  • Formula:
    C₁₅H₂₁NO·HCl
  • Storage Temperature:
    –20 °C
  • Description:
    α-Pyrrolidinopentiophenone (hydrochloride)
  • Cat. No.:
    76544-310

Specifications

About this item

A Certified Reference Material

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard