(±)-11-hydroxy-Δ9-THC (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
Certificates
About this item
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard
Specifications
- CAS No.:34675-49-5
- Synonym(s):(±)-11-hydroxy-Δ9-Tetrahydrocannabinol
- Formula:C₂₁H₃₀O₃
- Storage Temperature:–20 °C
- Description:(±)-11-hydroxy-Δ9-THC