Order Entry
Puerto Rico
ContactUsLinkComponent
Meperidine-d4 (hydrochloride) (CRM), Cayman Chemical
Meperidine-d4 (hydrochloride) (CRM), Cayman Chemical
Catalog # 76544-256
CAS Number:  
Meperidine-d4 (hydrochloride) (CRM), Cayman Chemical
Catalog # 76544-256
Supplier Number:  21323-1
CAS Number:  

Specifications

  • Concentration:
    1 mg/ml
  • Matrix:
    Methanol
  • Pack type:
    Glass ampule
  • Description:
    Meperidine-d4 (hydrochloride) (CRM)
  • Size:
    1 mg
  • Synonym(s):
    Pethidine-d4
  • Formula:
    C₁₅H₁₇D₄NO₂·HCl
  • Storage Temperature:
    –20 °C
  • Description:
    Meperidine-d4 (hydrochloride)
  • Cat. No.:
    76544-256

Specifications

About this item

A Certified Reference Material

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard