Order Entry
Puerto Rico
ContactUsLinkComponent
Phytocannabinoid Mixture 10 (CRM), Cayman Chemical
Phytocannabinoid Mixture 10 (CRM), Cayman Chemical
Catalog # 76544-246
CAS Number:  
Phytocannabinoid Mixture 10 (CRM), Cayman Chemical
Catalog # 76544-246
Supplier Number:  21305-1
CAS Number:  

Specifications

  • Concentration:
    250 µg/ml of each compound
  • Matrix:
    Acetonitrile
  • Pack type:
    Glass ampule
  • Description:
    Phytocannabinoid Mixture 10 (CRM)
  • Size:
    1 mg
  • Storage Temperature:
    –20 °C
  • Cat. No.:
    76544-246

Specifications

About this item

A Certified Reference Material cintaining a mixture of phytocannabinoids in an acetonitrile solution intended for HPLC-UV or LC- or GC-MS applications

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard