Order Entry
Puerto Rico
ContactUsLinkComponent
(±)-11-nor-9-carboxy-Δ9-THC (CRM), Cayman Chemical
(±)-11-nor-9-carboxy-Δ9-THC (CRM), Cayman Chemical
  76544-214
 :  
(±)-11-nor-9-carboxy-Δ9-THC (CRM), Cayman Chemical
  76544-214
 :  20754-1
 :  

 

  • Concentration:
    1 mg/ml
  • Matrix:
    Methanol
  • Pack type:
    Glass ampule
  • CAS No.:
    104874-50-2
  • Description:
    (±)-11-nor-9-carboxy-Δ9-THC (CRM)
  • Size:
    1 mg
  • Synonym(s):
    (±)-11-nor-9-carboxy-Δ9-Tetrahydrocannabinol
  • Formula:
    C₂₁H₂₈O₄
  • Storage Temperature:
    –20 °C
  • Description:
    (±)-11-nor-9-carboxy-Δ9-THC
  • Cat. No.:
    76544-214

 

 

A Certified Reference Material

  • This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
  • This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard