3,4-MDMA-d5 (hydrochloride) (CRM), Cayman Chemical
Supplier: Cayman Chemical Company
Certificates
About this item
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard
Specifications
- Synonym(s):3,4-Methylenedioxymethylamphetamine-d5
- Formula:C₁₁H₁₀D₅NO₂·HCl
- Storage Temperature:–20 °C
- Description:3,4-MDMA-d5 (hydrochloride)