Zaleplon-d4 (CRM), Cayman Chemical
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard
- CAS No.:1781887-91-9
- Formula:C₁₇H₁₁D₄N₅O
- Storage Temperature:–20 °C
- Description:Zaleplon-d4