Zopiclone (CRM), Cayman Chemical
76544-192
:
- Concentration:1 mg/ml
- Matrix:Acetonitrile
- Pack type:Glass ampule
- CAS No.:43200-80-2
- Description:Zopiclone (CRM)
- Size:1 mg
- Synonym(s):RP 27267; (±)-Zopiclone
- Formula:C₁₇H₁₇ClN₆O₃
- Storage Temperature:–20 °C
- Description:Zopiclone
- Cat. No.:76544-192
A Certified Reference Material
- This product is qualified as a Certified Reference Material (CRM) that has been manufactured and tested to meet ISO/IEC 17025 and ISO 17034 international standards
- This CRM has been characterized by metrologically valid procedures and may be used as a quantitative analytical reference standard