Order Entry
Puerto Rico
ContactUsLinkComponent
Zolpidem-d6 (CRM), Cayman Chemical
Zolpidem-d6 (CRM), Cayman Chemical
Certificates
Zolpidem-d6 (CRM), Cayman Chemical

Specifications

  • CAS No.:
    959605-90-4
  • Synonym(s):
    SL 80-0750-d6
  • Formula:
    C₁₉H₁₅D₆N₃O
  • Storage Temperature:
    –20 °C
  • Description:
    Zolpidem-d6

Specifications